missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442380050

 View more versions of this product

Catalog No. AC442380050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride
1.833
5g
96% min. (GC)
4-bromo-3-trifluoromethyl benzene sulphonyl chloride, 4-bromo-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-bromo-3-trifluoromethyl benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl benzenesulfonylchloride, 4-bromo-3-trifluoromethyl benzenesulphonyl chloride, 4-bromo-3-trifluoromethyl benzenesulphonylchloride, 4-bromo-3-trifluoromethyl phenyl chlorosulfone, 4-bromo-3-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl, bromo-3-trifluoromethyl benzenesulfonylchloride
C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Br
323.52
323.52
1.8330g/mL
30.0°C to 33.0°C
351003-47-9
C7H3BrClF3O2S
QCFIMBHKZZLZAE-UHFFFAOYSA-N
4-bromo-3-(trifluoromethyl)benzenesulfonyl chloride
2778606
97%

Chemical Identifiers

351003-47-9
323.52
4-bromo-3-trifluoromethyl benzene sulphonyl chloride, 4-bromo-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-bromo-3-trifluoromethyl benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl benzenesulfonylchloride, 4-bromo-3-trifluoromethyl benzenesulphonyl chloride, 4-bromo-3-trifluoromethyl benzenesulphonylchloride, 4-bromo-3-trifluoromethyl phenyl chlorosulfone, 4-bromo-3-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl, bromo-3-trifluoromethyl benzenesulfonylchloride
4-bromo-3-(trifluoromethyl)benzenesulfonyl chloride
C7H3BrClF3O2S
QCFIMBHKZZLZAE-UHFFFAOYSA-N
2778606
C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Br
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS Documents