missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)benzenesulfonamide, 97%

$66.62 - $252.91

Chemical Identifiers

CAS 351003-64-0
Molecular Formula C7H5BrF3NO2S
Molecular Weight (g/mol) 304.081
MDL Number MFCD03094285
InChI Key GQUUOLHRLZCAIO-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl benzene sulfonamide
PubChem CID 2778389
IUPAC Name 4-bromo-3-(trifluoromethyl)benzenesulfonamide
SMILES C1=CC(=C(C=C1S(=O)(=O)N)C(F)(F)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1970903 View Documents Alfa Aesar™
L1970903
1g Each for $66.62
 
AAL1970906 View Documents Alfa Aesar™
L1970906
5g Each for $252.91
 
Specifications

Chemical Identifiers

351003-64-0
304.081
GQUUOLHRLZCAIO-UHFFFAOYSA-N
2778389
C1=CC(=C(C=C1S(=O)(=O)N)C(F)(F)F)Br
C7H5BrF3NO2S
MFCD03094285
4-bromo-3-trifluoromethyl benzene sulfonamide
4-bromo-3-(trifluoromethyl)benzenesulfonamide

Specifications

185°C to 189°C
351003-64-0
MFCD03094285
GQUUOLHRLZCAIO-UHFFFAOYSA-N
4-bromo-3-(trifluoromethyl)benzenesulfonamide
2778389
97%
1g
C7H5BrF3NO2S
4-bromo-3-trifluoromethyl benzene sulfonamide
C1=CC(=C(C=C1S(=O)(=O)N)C(F)(F)F)Br
304.081
304.1
4-Bromo-3-(trifluoromethyl)benzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning