Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)aniline, 98%

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-(trifluoromethyl)aniline
CAS 393-36-2
Molecular Formula C7H5BrF3N
Formula Weight 240.03
MDL Number MFCD00007827
UN Number UN2811
Beilstein 641589
Synonym 5-Amino-2-bromobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1838003

  SDS
alfa aesar™
A1838003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1838006

  SDS
alfa aesar™
A1838006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.03
Health Hazard 1 Danger
Assay 98%
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Boiling Point 81° to 84°C (0.5mmHg)
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)aniline
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
UN Number UN2811
Flash Point >110°C (230°F)
Molecular Formula C7H5BrF3N
Synonym 5-Amino-2-bromobenzotrifluoride
MDL Number MFCD00007827
Melting Point 46° to 49°C
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
CAS 393-36-2
Beilstein 641589
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.