missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Bromo-3'-(trifluoromethyl)acetanilide, 97%

$78.04 - $78.04

Chemical Identifiers

CAS 41513-05-7
Molecular Formula C9H7BrF3NO
Formula Weight 282.07
MDL Number MFCD00052361
Synonym 5-Acetamido-2-bromobenzotrifluoride
Chemical Name or Material 4'-Bromo-3'-(trifluoromethyl)acetanilide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1993988 SDS Alfa Aesar™
A1993988
2.5g Each for $78.04
AAA1993909 SDS Alfa Aesar™
A1993909
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

41513-05-7
282.07
5-Acetamido-2-bromobenzotrifluoride
C9H7BrF3NO
MFCD00052361
4'-Bromo-3'-(trifluoromethyl)acetanilide

Specifications

41513-05-7
282.07
116° to 119°C
2.5g
97%
C9H7BrF3NO
MFCD00052361
5-Acetamido-2-bromobenzotrifluoride
4'-Bromo-3'-(trifluoromethyl)acetanilide

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning