Please login to your online account to display your discounted pricing

4-Bromo-3-phenyl-1H-pyrazol-5-amine, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-phenyl-1H-pyrazol-5-amine
CAS 2845-78-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H8BrN3
Formula Weight 238.09
MDL Number MFCD00067992
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM00411EA

  SDS
fisher scientific company
KM00411EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 238.09
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material 4-Bromo-3-phenyl-1H-pyrazol-5-amine
MDL Number MFCD00067992
CAS 2845-78-5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Assay Percent Range 95%
Molecular Formula C9H8BrN3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle