missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3-nitrotoluene, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1071814

 View more versions of this product

Catalog No. AAA1071814



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-3-nitrotoluene
29°C to 32°C
96°C (1 mmHg)
C7H6BrNO2
25 g
Light sensitive
UPBUTKQMDPHQAQ-UHFFFAOYSA-N
1-bromo-4-methyl-2-nitrobenzene
79224
≥98%
5326-34-1
1.578
>110°C (230°F)
MFCD00024180
1945925
4-bromo-3-nitrotoluene, 2-bromo-5-methylnitrobenzene, benzene, 1-bromo-4-methyl-2-nitro, bromonitrotoluene-4, 1-bromo-4-methyl-2-nitro-benzene, 3-nitro-4-bromotoluene, pubchem9585, acmc-209l5d, ksc490m7t, tpc-i121
CC1=CC=C(Br)C(=C1)[N+]([O-])=O
216.03
216.04

Chemical Identifiers

5326-34-1
216.03
UPBUTKQMDPHQAQ-UHFFFAOYSA-N
79224
CC1=CC=C(Br)C(=C1)[N+]([O-])=O
C7H6BrNO2
MFCD00024180
4-bromo-3-nitrotoluene, 2-bromo-5-methylnitrobenzene, benzene, 1-bromo-4-methyl-2-nitro, bromonitrotoluene-4, 1-bromo-4-methyl-2-nitro-benzene, 3-nitro-4-bromotoluene, pubchem9585, acmc-209l5d, ksc490m7t, tpc-i121
1-bromo-4-methyl-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 226-203-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only