missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3-nitrobenzotrifluoride, TCI America™

$59.58 - $180.28

Chemical Identifiers

CAS 349-03-1
Molecular Formula C7H3BrF3NO2
Molecular Weight (g/mol) 270.005
MDL Number MFCD00014685
InChI Key PESPBNYBZVIGRO-UHFFFAOYSA-N
Synonym 4-bromo-3-nitrobenzotrifluoride, 1-bromo-2-nitro-4-trifluoromethyl benzene, 2-nitro-4-trifluoromethylbromobenzene, 3-nitro-4-bromobenzotrifluoride, 2-bromo-5-trifluoromethyl nitrobenzene, 4-bromo-3-nitro-a,a,a-trifluorotoluene, benzene, 1-bromo-2-nitro-4-trifluoromethyl, 1-bromo-2-nitro-4-trifluoromethyl-benzene, 2-bromo-1-nitro-5-trifluoromethyl benzene, toluene, 4-bromo-3-nitro-alpha,alpha,alpha-trifluoro
PubChem CID 520706
IUPAC Name 1-bromo-2-nitro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30745G View Documents TCI America
B30745G
5g Each for $59.58
 
B307425G View Documents TCI America
B307425G
25g Each for $180.28
 
Specifications

Chemical Identifiers

349-03-1
270.005
PESPBNYBZVIGRO-UHFFFAOYSA-N
520706
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])Br
C7H3BrF3NO2
MFCD00014685
4-bromo-3-nitrobenzotrifluoride, 1-bromo-2-nitro-4-trifluoromethyl benzene, 2-nitro-4-trifluoromethylbromobenzene, 3-nitro-4-bromobenzotrifluoride, 2-bromo-5-trifluoromethyl nitrobenzene, 4-bromo-3-nitro-a,a,a-trifluorotoluene, benzene, 1-bromo-2-nitro-4-trifluoromethyl, 1-bromo-2-nitro-4-trifluoromethyl-benzene, 2-bromo-1-nitro-5-trifluoromethyl benzene, toluene, 4-bromo-3-nitro-alpha,alpha,alpha-trifluoro
1-bromo-2-nitro-4-(trifluoromethyl)benzene

Specifications

105°C
5g
C7H3BrF3NO2
4-bromo-3-nitrobenzotrifluoride, 1-bromo-2-nitro-4-trifluoromethyl benzene, 2-nitro-4-trifluoromethylbromobenzene, 3-nitro-4-bromobenzotrifluoride, 2-bromo-5-trifluoromethyl nitrobenzene, 4-bromo-3-nitro-a,a,a-trifluorotoluene, benzene, 1-bromo-2-nitro-4-trifluoromethyl, 1-bromo-2-nitro-4-trifluoromethyl-benzene, 2-bromo-1-nitro-5-trifluoromethyl benzene, toluene, 4-bromo-3-nitro-alpha,alpha,alpha-trifluoro
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])Br
270.005
270.01
≥98.0% (GC)
Yellow
349-03-1
MFCD00014685
PESPBNYBZVIGRO-UHFFFAOYSA-N
1-bromo-2-nitro-4-(trifluoromethyl)benzene
520706
Clear Liquid at 20°C
4-Bromo-3-nitrobenzotrifluoride
Safety and Handling

Safety and Handling

TSCA : No