Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-nitrobenzotrifluoride, 98+%

$53.42 - $53.42

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-nitrobenzotrifluoride
CAS 349-03-1
Molecular Formula C7H3BrF3NO2
Formula Weight 270.01
MDL Number MFCD00014685
Beilstein 2417762
Synonym 1-Bromo-2-nitro-4-(trifluoromethyl)benzene; 2-Bromo-5-(trifluoromethyl)nitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2034806

 
alfa aesar™
B2034806
5g Each for $53.42
Description & Specifications

Specifications

Formula Weight 270.01
Chemical Name or Material 4-Bromo-3-nitrobenzotrifluoride
Assay ≥98%
Molecular Formula C7H3BrF3NO2
MDL Number MFCD00014685
CAS 349-03-1
Beilstein 2417762
Synonym 1-Bromo-2-nitro-4-(trifluoromethyl)benzene; 2-Bromo-5-(trifluoromethyl)nitrobenzene
Boiling Point 90° to 92°C (5mmHg)
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P337+P313
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338-P337+P313
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.