missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-3-methylbenzoic Acid 97.0+%, TCI America™
Supplier: TCI America B311325G
Specifications
4-Bromo-3-methylbenzoic Acid | |
215°C | |
C8H6BrO2 | |
25 g | |
KWVXDZLVCISXIB-UHFFFAOYSA-M | |
4-bromo-3-methylbenzoate | |
82131 | |
≥97.0% (GC,T) |
7697-28-1 | |
White-Yellow | |
MFCD00039526 | |
4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b | |
CC1=CC(=CC=C1Br)C([O-])=O | |
214.04 | |
215.05 | |
Crystalline Powder |
Chemical Identifiers
7697-28-1 | |
214.04 | |
KWVXDZLVCISXIB-UHFFFAOYSA-M | |
82131 | |
CC1=CC(=CC=C1Br)C([O-])=O |
C8H6BrO2 | |
MFCD00039526 | |
4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b | |
4-bromo-3-methylbenzoate |
Safety and Handling
TSCA : No