missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3-methylbenzenesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442390010

 View more versions of this product

Catalog No. AC442390010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Bromo-3-methylbenzenesulfonyl chloride
Glass Bottle
58°C to 60°C
72256-93-0
100.0
C7H6BrClO2S
JRNDXUKMWFVEIC-UHFFFAOYSA-N
4-bromo-3-methylbenzenesulfonyl chloride
2737503
Crystalline Powder
Authentic
Brown to White
1g
96.0
96% min. (GC)
4-bromo-3-methyl-benzenesulfonyl chloride, 4-bromo-3-methylbenzene sulfonyl chloride, 4-bromo-3-methylbenzene-1-sulfonyl chloride, 4-bromo-3-methylbenzenesulphonyl chloride, 4-bromo-3-methylbenzenesulphonylchloride, 4-bromo-3-methylphenyl chlorosulfone, 6-bromo-m-toluene-3-sulfonyl chloride, acmc-1bho4, jrndxukmwfveic-uhfffaoysa, pubchem15805
CC1=C(C=CC(=C1)S(=O)(=O)Cl)Br
269.55
269.55
97%

Chemical Identifiers

72256-93-0
269.55
4-bromo-3-methyl-benzenesulfonyl chloride, 4-bromo-3-methylbenzene sulfonyl chloride, 4-bromo-3-methylbenzene-1-sulfonyl chloride, 4-bromo-3-methylbenzenesulphonyl chloride, 4-bromo-3-methylbenzenesulphonylchloride, 4-bromo-3-methylphenyl chlorosulfone, 6-bromo-m-toluene-3-sulfonyl chloride, acmc-1bho4, jrndxukmwfveic-uhfffaoysa, pubchem15805
4-bromo-3-methylbenzenesulfonyl chloride
C7H6BrClO2S
JRNDXUKMWFVEIC-UHFFFAOYSA-N
2737503
CC1=C(C=CC(=C1)S(=O)(=O)Cl)Br
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents