missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3-methylaniline, 97%, Acros Organics™

Manufacturer:  Acros Organics AC167500500

 View more versions of this product

Catalog No. AC167500500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Bromo-3-methylaniline
6933-10-4
100.0
Glass bottle
240.0°C
Brown to White
97%
BrC6H3(CH3)NH2
12, 873
MMEGELSFOYDPQW-UHFFFAOYSA-N
4-bromo-3-methylaniline
23359
Powder
99%
98.5
Authentic
79.0°C to 83.0°C
50g
97%
C7H8BrN
MFCD00007828
4-Bromo-m-toluidine
CC1=C(C=CC(=C1)N)Br
186.05
186.05

Chemical Identifiers

6933-10-4
186.05
MMEGELSFOYDPQW-UHFFFAOYSA-N
23359
C7H8BrN
MFCD00007828
4-Bromo-m-toluidine
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 230-056-3

SDS Documents