Learn More
4-Bromo-3-methylaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1541709
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Bromo-3-methylaniline | |
79°C to 83°C | |
Characteristic | |
MFCD00007828 | |
UN2811 | |
4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline | |
CC1=C(C=CC(=C1)N)Br | |
186.05 | |
186.06 |
6933-10-4 | |
239°C to 241°C | |
C7H8BrN | |
10 g | |
2689600 | |
MMEGELSFOYDPQW-UHFFFAOYSA-N | |
4-bromo-3-methylaniline | |
23359 | |
97% |
Chemical Identifiers
6933-10-4 | |
186.05 | |
MMEGELSFOYDPQW-UHFFFAOYSA-N | |
23359 | |
CC1=C(C=CC(=C1)N)Br |
C7H8BrN | |
MFCD00007828 | |
4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline | |
4-bromo-3-methylaniline |
Safety and Handling
GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RUO – Research Use Only