Learn More
4-Bromo-3-methylaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1541730
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Bromo-3-methylaniline | |
79°C to 83°C | |
Characteristic | |
MFCD00007828 | |
UN2811 | |
4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline | |
CC1=C(C=CC(=C1)N)Br | |
186.05 | |
186.06 |
6933-10-4 | |
239°C to 241°C | |
C7H8BrN | |
250 g | |
2689600 | |
MMEGELSFOYDPQW-UHFFFAOYSA-N | |
4-bromo-3-methylaniline | |
23359 | |
97% |
Chemical Identifiers
6933-10-4 | |
186.05 | |
MMEGELSFOYDPQW-UHFFFAOYSA-N | |
23359 | |
CC1=C(C=CC(=C1)N)Br |
C7H8BrN | |
MFCD00007828 | |
4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline | |
4-bromo-3-methylaniline |
Safety and Handling
GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 230-056-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only