missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-methylaniline, 97%

Manufacturer:  Alfa Aesar™ A1541718

 View more versions of this product

Catalog No. AAA1541718



Specifications

Specifications

4-Bromo-3-methylaniline
239°C to 241°C
50g
C7H8BrN
UN2811
4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline
CC1=C(C=CC(=C1)N)Br
186.05
186.06
Characteristic
79°C to 83°C
6933-10-4
MFCD00007828
2689600
MMEGELSFOYDPQW-UHFFFAOYSA-N
4-bromo-3-methylaniline
23359
97%

Chemical Identifiers

6933-10-4
186.05
MMEGELSFOYDPQW-UHFFFAOYSA-N
23359
CC1=C(C=CC(=C1)N)Br
C7H8BrN
MFCD00007828
4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline
4-bromo-3-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents