missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-Bromo-3′-methylacetanilide, TCI America™

$44.31 - $113.06

Chemical Identifiers

CAS 90914-81-1
Molecular Formula C9H10BrNO
Molecular Weight (g/mol) 228.089
MDL Number MFCD00040822
InChI Key BYZHUFNLXFFINU-UHFFFAOYSA-N
Synonym n-4-bromo-3-methylphenyl acetamide, 4'-bromo-3'-methylacetanilide, 4-bromo-3-methylacetanilide, n-acetyl-4-bromo-m-toluidine, 3-methyl-4-bromoacetanilide, n-acetyl 4-bromo-3-methylaniline, n-4-bromo-3-methylphenyl-acetamide, n-4-bromo-3-methylphenyl-acetamide, maybridge1_005107, acmc-209r7e
PubChem CID 738336
IUPAC Name N-(4-bromo-3-methylphenyl)acetamide
SMILES CC1=C(C=CC(=C1)NC(=O)C)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B31425G View Documents TCI America
B31425G
5g Each for $44.31
 
B314225G View Documents TCI America
B314225G
25g Each for $113.06
 
Specifications

Chemical Identifiers

90914-81-1
228.089
BYZHUFNLXFFINU-UHFFFAOYSA-N
738336
CC1=C(C=CC(=C1)NC(=O)C)Br
C9H10BrNO
MFCD00040822
n-4-bromo-3-methylphenyl acetamide, 4'-bromo-3'-methylacetanilide, 4-bromo-3-methylacetanilide, n-acetyl-4-bromo-m-toluidine, 3-methyl-4-bromoacetanilide, n-acetyl 4-bromo-3-methylaniline, n-4-bromo-3-methylphenyl-acetamide, n-4-bromo-3-methylphenyl-acetamide, maybridge1_005107, acmc-209r7e
N-(4-bromo-3-methylphenyl)acetamide

Specifications

Undesignated
5g
C9H10BrNO
n-4-bromo-3-methylphenyl acetamide, 4'-bromo-3'-methylacetanilide, 4-bromo-3-methylacetanilide, n-acetyl-4-bromo-m-toluidine, 3-methyl-4-bromoacetanilide, n-acetyl 4-bromo-3-methylaniline, n-4-bromo-3-methylphenyl-acetamide, n-4-bromo-3-methylphenyl-acetamide, maybridge1_005107, acmc-209r7e
CC1=C(C=CC(=C1)NC(=O)C)Br
228.089
228.09
≥98.0% (GC)
103°C
90914-81-1
MFCD00040822
BYZHUFNLXFFINU-UHFFFAOYSA-N
N-(4-bromo-3-methylphenyl)acetamide
738336
Crystal-Powder at 20°C
4′-Bromo-3′-methylacetanilide
Safety and Handling

Safety and Handling

TSCA : No