Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Bromo-3'-methylacetanilide, 98%

$33.15 - $328.47

Chemical Identifiers

Chemical Name or Material 4'-Bromo-3'-methylacetanilide
CAS 90914-81-1
Molecular Formula C9H10BrNO
Formula Weight 228.09
MDL Number MFCD00040822
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2198022

  SDS
alfa aesar™
B2198022
100g Each for $328.47

AAB2198006

  SDS
alfa aesar™
B2198006
5g Each for $33.15
Description & Specifications

Specifications

Formula Weight 228.09
Chemical Name or Material 4'-Bromo-3'-methylacetanilide
Assay 98%
Molecular Formula C9H10BrNO
MDL Number MFCD00040822
CAS 90914-81-1
Melting Point 102° to 104°C
Quantity 100g