Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-methyl-5-trifluoromethyl-1H-pyrazole, 98%

$212.42 - $949.73

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-methyl-5-trifluoromethyl-1H-pyrazole
CAS 60061-68-9
Molecular Formula C5H4BrF3N2
Formula Weight 229
MDL Number MFCD08060951
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5065703

  SDS
alfa aesar™
H5065703
1g Each for $212.42

AAH5065706

  SDS
alfa aesar™
H5065706
5g Each for $949.73
Description & Specifications

Specifications

Formula Weight 229
Chemical Name or Material 4-Bromo-3-methyl-5-trifluoromethyl-1H-pyrazole
Assay 98%
Molecular Formula C5H4BrF3N2
MDL Number MFCD08060951
CAS 60061-68-9
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.