Please login to your online account to display your discounted pricing

4-Bromo-3-fluorotoluene, 98%, ACROS Organics™

$109.67 - $109.67

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-fluorotoluene
CAS 452-74-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4BrFO
Linear Formula CH3C6H3(Br)F
Formula Weight 191.01
MDL Number MFCD00040940
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC311090250

  SDS
thermo fisher scientific
AC311090250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC311090050

  SDS
thermo fisher scientific
311090050
Glass bottle 5g Each for $109.67
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 191.01
Quantity 25g
Color Clear light yellow
Density 1.4900g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C6H4BrFO
Assay Percent Range 98%
CAS 452-74-4
Chemical Name or Material 4-Bromo-3-fluorotoluene
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
MDL Number MFCD00040940
CAS Min % 97.5
Refractive Index 1.5290 to 1.5310 (20°C, 589nm)
Linear Formula CH3C6H3(Br)F
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.