Learn More
4-Bromo-3-fluorotoluene, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 311090050
Specifications
4-Bromo-3-fluorotoluene | |
97.5 | |
Yellow | |
95.0°C (50.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5290 to 1.5310 | |
MFCD00040940 | |
1.49 | |
SLFNGVGRINFJLK-UHFFFAOYSA-N | |
1-bromo-2-fluoro-4-methylbenzene | |
573280 | |
98% |
452-74-4 | |
100.0 | |
1.4900g/mL | |
34°C | |
98% | |
C6H4BrFO | |
CH3C6H3(Br)F | |
5 g | |
4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562 | |
CC1=CC(=C(C=C1)Br)F | |
191.01 | |
191.01 | |
Liquid |
Chemical Identifiers
452-74-4 | |
SLFNGVGRINFJLK-UHFFFAOYSA-N | |
573280 | |
CC1=CC(=C(C=C1)Br)F |
MFCD00040940 | |
4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562 | |
1-bromo-2-fluoro-4-methylbenzene |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only