Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-fluorobenzotrifluoride, 97%

$59.38 - $212.46

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-fluorobenzotrifluoride
CAS 40161-54-4
Molecular Formula C7H3BrF4
Formula Weight 243
MDL Number MFCD00042497
Beilstein 2641902
Synonym 1-Bromo-2-fluoro-4-(trifluoromethyl)benzene; 2-Fluoro-4-(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2011703

  SDS
alfa aesar™
B2011703
1g Each for $59.38

AAB2011706

  SDS
alfa aesar™
B2011706
5g Each for $212.46
Description & Specifications

Specifications

Formula Weight 243
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2641902
Boiling Point 154° to 155°C
Molecular Formula C7H3BrF4
Assay 97%
CAS 40161-54-4
MDL Number MFCD00042497
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 1-Bromo-2-fluoro-4-(trifluoromethyl)benzene; 2-Fluoro-4-(trifluoromethyl)bromobenzene
Chemical Name or Material 4-Bromo-3-fluorobenzotrifluoride
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.