missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3-fluorobenzenesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442360010

 View more versions of this product

Catalog No. AC442360010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Bromo-3-fluorobenzenesulfonyl chloride
Glass Bottle
Colorless
351003-51-5
100.0
C6H3BrClFO2S
IZCXVIACMHTZNA-UHFFFAOYSA-N
4-bromo-3-fluorobenzenesulfonyl chloride
2778218
Liquid
Authentic
130.0°C (1.0 mmHg)
1g
96.0
96% min. (GC)
3-fluoro-4-bromobenzenesulfonyl chloride, 4-bromo-3-fluoro-benzenesulfonyl chloride, 4-bromo-3-fluorobenzene sulfonyl chloride, 4-bromo-3-fluorobenzene-1-sulfonyl chloride, 4-bromo-3-fluorobenzenesulfonylchloride, 4-bromo-3-fluorobenzenesulphonyl chloride, acmc-209vnt, benzenesulfonyl chloride, 4-bromo-3-fluoro, buttpark 76\07-90, pubchem2220
C1=CC(=C(C=C1S(=O)(=O)Cl)F)Br
273.51
273.51
97%

Chemical Identifiers

351003-51-5
273.51
3-fluoro-4-bromobenzenesulfonyl chloride, 4-bromo-3-fluoro-benzenesulfonyl chloride, 4-bromo-3-fluorobenzene sulfonyl chloride, 4-bromo-3-fluorobenzene-1-sulfonyl chloride, 4-bromo-3-fluorobenzenesulfonylchloride, 4-bromo-3-fluorobenzenesulphonyl chloride, acmc-209vnt, benzenesulfonyl chloride, 4-bromo-3-fluoro, buttpark 76\07-90, pubchem2220
4-bromo-3-fluorobenzenesulfonyl chloride
C6H3BrClFO2S
IZCXVIACMHTZNA-UHFFFAOYSA-N
2778218
C1=CC(=C(C=C1S(=O)(=O)Cl)F)Br
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Protect from moisture.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents