Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-fluorobenzenesulfonamide, 97%

$109.54 - $423.41

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-fluorobenzenesulfonamide
CAS 263349-73-1
Molecular Formula C6H5BrFNO2S
Formula Weight 254.08
MDL Number MFCD03094292
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1970703

  SDS
alfa aesar™
L1970703
1g Each for $109.54

AAL1970706

  SDS
alfa aesar™
L1970706
5g Each for $423.41
Description & Specifications

Specifications

Formula Weight 254.08
Chemical Name or Material 4-Bromo-3-fluorobenzenesulfonamide
MDL Number MFCD03094292
CAS 263349-73-1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Quantity 1g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 137° to 141°C
Molecular Formula C6H5BrFNO2S
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.