Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-fluoroanisole, 97%

$67.05 - $641.88

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-fluoroanisole
CAS 458-50-4
Molecular Formula C7H6BrFO
Formula Weight 205.03
MDL Number MFCD01310983
Synonym 1-Bromo-2-fluoro-4-methoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2731903

  SDS
alfa aesar™
H2731903
1g Each for $67.05

AAH2731906

  SDS
alfa aesar™
H2731906
5g Each for $186.84

AAH2731914

  SDS
alfa aesar™
H2731914
25g Each for $641.88
Description & Specifications

Specifications

Formula Weight 205.03
Synonym 1-Bromo-2-fluoro-4-methoxybenzene
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Bromo-3-fluoroanisole
Melting Point 217° to 219°C
Assay 97%
Quantity 1g
Flash Point 85°C (185°F)
Molecular Formula C7H6BrFO
MDL Number MFCD01310983
CAS 458-50-4
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.