Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-fluoroaniline, 98%

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-fluoroaniline
CAS 656-65-5
Molecular Formula C6H5BrFN
Formula Weight 190.01
MDL Number MFCD00672933
UN Number UN2811
Beilstein 3236457
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2222709

 
alfa aesar™
B2222709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.01
Chemical Name or Material 4-Bromo-3-fluoroaniline
Melting Point 65° to 70°C
Assay 98%
Quantity 10g
Flash Point 104°C (219°F)
Molecular Formula C6H5BrFN
MDL Number MFCD00672933
CAS 656-65-5
UN Number UN2811
Beilstein 3236457
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Applications
4-Bromo-3-fluoroaniline is an intermediate used for the synthetic preparation of Tradizolid and various pharmaceuticals such as the synthesis of several lateral difluoro-substituted 4,4″-dialkyl- and 4,4″-alkoxyalkylterphenyls.

Solubility
Soluble in acetonitrile, methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.