Please login to your online account to display your discounted pricing

Alfa Aesar™ Alfa Aesar™ 4'-Bromo-3'-fluoroacetanilide, 98+%

$72.32 - $223.00

Chemical Identifiers

Chemical Name or Material 4'-Bromo-3'-fluoroacetanilide
CAS 351-30-4
Molecular Formula C8H7BrFNO
Formula Weight 232.05
MDL Number MFCD00672937
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2553109

  SDS
alfa aesar™
B2553109
10g Each for $72.32

AAB2553118

  SDS
alfa aesar™
B2553118
50g Each for $223.00
Description & Specifications

Specifications

Formula Weight 232.05
Health Hazard 1 Warning
MDL Number MFCD00672937
Molecular Formula C8H7BrFNO
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 149° to 152°C
CAS 351-30-4
Quantity 10g
Chemical Name or Material 4'-Bromo-3'-fluoroacetanilide
Assay ≥98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.