Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-ethoxyaniline hydrochloride, 98%

$45.05 - $473.12

Chemical Identifiers

Chemical Name or Material 4-Bromo-3-ethoxyaniline hydrochloride
CAS 846023-33-4
Molecular Formula C8H10BrNO·HCl
Formula Weight 216.08
MDL Number MFCD03790891
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2482706

  SDS
alfa aesar™
B2482706
5g Each for $45.05

AAB2482722

  SDS
alfa aesar™
B2482722
100g Each for $473.12
Description & Specifications

Specifications

Formula Weight 216.08
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Melting Point 186° to 189°C
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 846023-33-4
Molecular Formula C8H10BrNO·HCl
Chemical Name or Material 4-Bromo-3-ethoxyaniline hydrochloride
Quantity 5g
MDL Number MFCD03790891
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.