missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3-chlorobenzotrifluoride, 98%

Manufacturer:  Alfa Aesar™ H6197822

 View more versions of this product

Catalog No. AAH6197822



Specifications

Specifications

4-Bromo-3-chlorobenzotrifluoride
402-04-0
MFCD00673985
RVTIHGGJJMXISV-UHFFFAOYSA-N
1-bromo-2-chloro-4-(trifluoromethyl)benzene
14273266
Liquid
100g
C7H3BrClF3
1-bromo-2-chloro-4-trifluoromethyl benzene
C1=CC(=C(C=C1C(F)(F)F)Cl)Br
259.45
259.45
98%

Chemical Identifiers

402-04-0
259.45
RVTIHGGJJMXISV-UHFFFAOYSA-N
14273266
C1=CC(=C(C=C1C(F)(F)F)Cl)Br
C7H3BrClF3
MFCD00673985
1-bromo-2-chloro-4-trifluoromethyl benzene
1-bromo-2-chloro-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents