missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-Bromo-3′-chloroacetanilide, TCI America™

$71.81 - $220.00

Chemical Identifiers

CAS 22459-81-0
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.504
MDL Number MFCD00040848
InChI Key BVADKOYGDKTWMU-UHFFFAOYSA-N
Synonym 4'-bromo-3'-chloroacetanilide, n-4-bromo-3-chlorophenyl acetamide, n-acetyl 4-bromo-3-chloroaniline, 4-bromo-3-chloroacetanilide, n1-4-bromo-3-chlorophenyl acetamide, acetamide,n-4-bromo-3-chlorophenyl, acmc-209fwl, maybridge1_008946, n-acetyl4-bromo-3-chloroaniline
PubChem CID 2746295
IUPAC Name N-(4-bromo-3-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B31415G View Documents TCI America
B31415G
5g Each for $71.81
 
B314125G View Documents TCI America
B314125G
25g Each for $220.00
 
Specifications

Chemical Identifiers

22459-81-0
248.504
BVADKOYGDKTWMU-UHFFFAOYSA-N
2746295
CC(=O)NC1=CC(=C(C=C1)Br)Cl
C8H7BrClNO
MFCD00040848
4'-bromo-3'-chloroacetanilide, n-4-bromo-3-chlorophenyl acetamide, n-acetyl 4-bromo-3-chloroaniline, 4-bromo-3-chloroacetanilide, n1-4-bromo-3-chlorophenyl acetamide, acetamide,n-4-bromo-3-chlorophenyl, acmc-209fwl, maybridge1_008946, n-acetyl4-bromo-3-chloroaniline
N-(4-bromo-3-chlorophenyl)acetamide

Specifications

White-Yellow
5g
C8H7BrClNO
4'-bromo-3'-chloroacetanilide, n-4-bromo-3-chlorophenyl acetamide, n-acetyl 4-bromo-3-chloroaniline, 4-bromo-3-chloroacetanilide, n1-4-bromo-3-chlorophenyl acetamide, acetamide,n-4-bromo-3-chlorophenyl, acmc-209fwl, maybridge1_008946, n-acetyl4-bromo-3-chloroaniline
CC(=O)NC1=CC(=C(C=C1)Br)Cl
248.504
248.50
≥98.0% (GC)
125°C
22459-81-0
MFCD00040848
BVADKOYGDKTWMU-UHFFFAOYSA-N
N-(4-bromo-3-chlorophenyl)acetamide
2746295
Crystal-Powder at 20°C
4′-Bromo-3′-chloroacetanilide
Safety and Handling

Safety and Handling

TSCA : No