Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Bromo-3'-chloroacetanilide, 98%

$40.69 - $40.69

Chemical Identifiers

Chemical Name or Material 4'-Bromo-3'-chloroacetanilide
CAS 22459-81-0
Molecular Formula C8H7BrClNO
Formula Weight 248.51
MDL Number MFCD00040848
Beilstein 2967800
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1654706

 
alfa aesar™
A1654706
5g Each for $40.69
Description & Specifications

Specifications

Formula Weight 248.51
Chemical Name or Material 4'-Bromo-3'-chloroacetanilide
Assay 98%
Molecular Formula C8H7BrClNO
MDL Number MFCD00040848
CAS 22459-81-0
Melting Point 123° to 125°C
Beilstein 2967800
Quantity 5g