missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3,5-dimethylphenol 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC106871000

 View more versions of this product

Catalog No. AC106871000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Bromo-3, 5-dimethylphenol
7463-51-6
100.0
Glass bottle
≥98.5%
C8H9BrO
201.06
Colorless to slightly pink
113° to 116°C
99%
98.5
Authentic
Crystals or Powder
99%
BrC6H2(CH3)2OH
MFCD00002315
4-Bromo-3, 5-xylenol
100g

Chemical Identifiers

7463-51-6
100.0
C8H9BrO
201.06
4-Bromo-3, 5-xylenol
≥98.5%
98.5
99%
BrC6H2(CH3)2OH
MFCD00002315
4-Bromo-3, 5-dimethylphenol, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 231-255-8