Please login to your online account to display your discounted pricing

4-Bromo-3,5-dimethylisoxazole, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC00610DE

 View more versions of this product

Catalog No. CC00610DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 176.01
Quantity 5g
Assay 97%
Density 1.484g/mL
CAS Max % 100.0
Molecular Formula C5H6BrNO
CAS Min % 96.5
MDL Number MFCD00068187
Assay Percent Range 97%
CAS 10558-25-5
Chemical Name or Material 4-Bromo-3, 5-dimethylisoxazole

Specifications

Chemical Name or Material 4-Bromo-3, 5-dimethylisoxazole
CAS 10558-25-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6BrNO
Formula Weight 176.01
MDL Number MFCD00068187