missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3,5-dimethyl-1H-pyrazole, 99%

Manufacturer:  Alfa Aesar™ A1175914

 View more versions of this product

Catalog No. AAA1175914



Specifications

Specifications

4-Bromo-3,5-dimethyl-1H-pyrazole
25g
C5H7BrN2
108534
RISOHYOEPYWKOB-UHFFFAOYSA-N
4-bromo-3,5-dimethyl-1H-pyrazole
76937
99%
118°C to 123°C
3398-16-1
MFCD00005242
4-bromo-3,5-dimethylpyrazole, pyrazole, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl, 3.5-dimethyl-4-bromopyrazole, acmc-1cnce, 5-23-05-00154 beilstein handbook reference, ksc495g9b, 4-bromo-3,5-dimethyl pyrazole, 4-bromo-3,5-dimethyl-pyrazole
CC1=C(C(=NN1)C)Br
175.029
175.03

Chemical Identifiers

3398-16-1
175.029
RISOHYOEPYWKOB-UHFFFAOYSA-N
76937
CC1=C(C(=NN1)C)Br
C5H7BrN2
MFCD00005242
4-bromo-3,5-dimethylpyrazole, pyrazole, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl, 3.5-dimethyl-4-bromopyrazole, acmc-1cnce, 5-23-05-00154 beilstein handbook reference, ksc495g9b, 4-bromo-3,5-dimethyl pyrazole, 4-bromo-3,5-dimethyl-pyrazole
4-bromo-3,5-dimethyl-1H-pyrazole
Safety and Handling

Safety and Handling

RTECSNumber : UQ6210000

SDS Documents