Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3,5-dihydroxybenzamide, 97%

$341.72 - $341.72

Chemical Identifiers

Chemical Name or Material 4-Bromo-3,5-dihydroxybenzamide
CAS 13429-12-4
Molecular Formula C7H6BrNO3
Formula Weight 232.03
MDL Number MFCD00017131
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2049318

  SDS
alfa aesar™
B2049318
50g Each for $341.72
Description & Specifications

Specifications

Formula Weight 232.03
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Bromo-3,5-dihydroxybenzamide
TSCA TSCA
Assay 97%
Molecular Formula C7H6BrNO3
MDL Number MFCD00017131
CAS 13429-12-4
Quantity 50g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.