Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3,5-difluorophenol, 99%

$170.86 - $170.86

Chemical Identifiers

Chemical Name or Material 4-Bromo-3,5-difluorophenol
CAS 130191-91-2
Molecular Formula C6H3BrF2O
Formula Weight 208.99
MDL Number MFCD08062385
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2615303

  SDS
alfa aesar™
H2615303
1g Each for $170.86
Description & Specifications

Specifications

Formula Weight 208.99
Chemical Name or Material 4-Bromo-3,5-difluorophenol
Assay 99%
Molecular Formula C6H3BrF2O
MDL Number MFCD08062385
CAS 130191-91-2
Melting Point 72° to 78°C
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
4-Bromo-3,5-difluorophenol is used as intermediates of Liquid Crystals. Reactant in the synthesis of difluoroethyleneoxy group liquid crystal.

Solubility
Sparingly Soluble (8.2E-5 g/L) (25°C).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.