Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3,5-difluoroanisole, 98%

$81.43 - $81.43

Chemical Identifiers

Chemical Name or Material 4-Bromo-3,5-difluoroanisole
CAS 202865-61-0
Molecular Formula C7H5BrF2O
Formula Weight 223.02
MDL Number MFCD00143268
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1742706

  SDS
alfa aesar™
A1742706
5g Each for $81.43
Description & Specifications

Specifications

Formula Weight 223.02
Chemical Name or Material 4-Bromo-3,5-difluoroanisole
Assay 98%
Molecular Formula C7H5BrF2O
MDL Number MFCD00143268
CAS 202865-61-0
Melting Point 26° to 30°C
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
It is used to prepare difluorophenacyl analogs as inhibitors of cyclin-dependent kinases. It is also used to synthesize aminopyridine N-oxides for selective inhibition of p38 MAP kinase.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.