Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3,5-dichlorobenzotrifluoride, 97%

$46.85 - $123.24

Chemical Identifiers

Chemical Name or Material 4-Bromo-3,5-dichlorobenzotrifluoride
CAS 118754-53-3
Molecular Formula C7H2BrCl2F3
Formula Weight 293.9
MDL Number MFCD00221020
Synonym 2-Bromo-1,3-dichloro-5-(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2858003

  SDS
alfa aesar™
H2858003
1g Each for $123.24

AAH28580MD

  SDS
alfa aesar™
H28580MD
250mg Each for $46.85
Description & Specifications

Specifications

Formula Weight 293.9
Chemical Name or Material 4-Bromo-3,5-dichlorobenzotrifluoride
Assay 97%
Molecular Formula C7H2BrCl2F3
MDL Number MFCD00221020
CAS 118754-53-3
Synonym 2-Bromo-1,3-dichloro-5-(trifluoromethyl)benzene
Boiling Point 91° to 92°C (10mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.