missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-3,3,4,4-tetrafluoro-1-butene, 98%

$66.93 - $200.47

Chemical Identifiers

CAS 18599-22-9
Molecular Formula C4H3BrF4
Molecular Weight (g/mol) 206.966
MDL Number MFCD00042189
InChI Key GVCWGFZDSIWLMO-UHFFFAOYSA-N
Synonym 4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene
PubChem CID 87721
IUPAC Name 4-bromo-3,3,4,4-tetrafluorobut-1-ene
SMILES C=CC(C(F)(F)Br)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1849206 View Documents Alfa Aesar™
L1849206
5g Each for $200.47
 
AAL1849203 View Documents Alfa Aesar™
L1849203
1g Each for $66.93
 
Specifications

Chemical Identifiers

18599-22-9
206.966
GVCWGFZDSIWLMO-UHFFFAOYSA-N
87721
C=CC(C(F)(F)Br)(F)F
C4H3BrF4
MFCD00042189
4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene
4-bromo-3,3,4,4-tetrafluorobut-1-ene

Specifications

55°C to 57°C
18599-22-9
BrCF2CF2CH=CH2
1856181
GVCWGFZDSIWLMO-UHFFFAOYSA-N
4-bromo-3,3,4,4-tetrafluorobut-1-ene
87721
98%
5g
C4H3BrF4
MFCD00042189
4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene
C=CC(C(F)(F)Br)(F)F
206.966
206.97
4-Bromo-3,3,4,4-tetrafluoro-1-butene
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA