missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-(trifluoromethyl)phenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1105103

 View more versions of this product

Catalog No. AAL1105103



Specifications

Specifications

4-Bromo-2-(trifluoromethyl)phenyl isothiocyanate
1g
C8H3BrF3NS
UN2922
4-bromo-2-trifluoromethyl phenyl isothiocyanate, 4-bromo-2-trifluoromethylphenylisothiocyanate, 4-bromo-2-trifluoromethylphenyl isothiocyanate, 4-bromo-1-isothiocyanato-2-trifluoromethyl benzene, 4-bromo-2-trifluoromethyl phenylisothiocyanate, acmc-20amnc, pubchem5030, 4-bromo-2-trifluoromethyl benzenisothiocyanate, 4-bromo-1-isothiocyanato-2-trifluoromethyl benzene #
C1=CC(=C(C=C1Br)C(F)(F)F)N=C=S
282.078
282.09
129°C to 130°C (7mmHg)
206559-46-8
MFCD00041086
6923715
GWLIKBAQYRDIID-UHFFFAOYSA-N
4-bromo-1-isothiocyanato-2-(trifluoromethyl)benzene
624649
97%

Chemical Identifiers

206559-46-8
282.078
GWLIKBAQYRDIID-UHFFFAOYSA-N
624649
C1=CC(=C(C=C1Br)C(F)(F)F)N=C=S
C8H3BrF3NS
MFCD00041086
4-bromo-2-trifluoromethyl phenyl isothiocyanate, 4-bromo-2-trifluoromethylphenylisothiocyanate, 4-bromo-2-trifluoromethylphenyl isothiocyanate, 4-bromo-1-isothiocyanato-2-trifluoromethyl benzene, 4-bromo-2-trifluoromethyl phenylisothiocyanate, acmc-20amnc, pubchem5030, 4-bromo-2-trifluoromethyl benzenisothiocyanate, 4-bromo-1-isothiocyanato-2-trifluoromethyl benzene #
4-bromo-1-isothiocyanato-2-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents