Please login to your online account to display your discounted pricing

4-Bromo-2-(trifluoromethyl)phenol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-(trifluoromethyl)phenol
CAS 50824-04-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4BrF3O
Formula Weight 241.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC438230010

  SDS
thermo fisher scientific
AC438230010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 241.01
Quantity 1g
Color White
Melting Point 84° to 88°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Infrared Spectrum Authentic
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)phenol
Molecular Formula C7H4BrF3O
CAS 50824-04-9
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.