Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-(trifluoromethyl)benzaldehyde, 95%

$138.62 - $142.24

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-(trifluoromethyl)benzaldehyde
CAS 861928-27-0
Molecular Formula C8H4BrF3O
Formula Weight 253.02
MDL Number MFCD09037472
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2698603

  SDS
alfa aesar™
H2698603
1g Each for $138.62

AAH26986MD

  SDS
alfa aesar™
H26986MD
250mg Each for $142.24
Description & Specifications

Specifications

Formula Weight 253.02
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)benzaldehyde
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H4BrF3O
MDL Number MFCD09037472
Assay 95%
CAS 861928-27-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
4-Bromo-2-(trifluoromethyl)benzaldehyde is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical intermediate and intermediate in organic syntheses.

Solubility
Sparingly soluble in water (0.083 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.