Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-(trifluoromethyl)anisole, 97%

$107.30 - $357.11

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-(trifluoromethyl)anisole
CAS 1514-11-0
Molecular Formula C8H6BrF3O
Formula Weight 255.04
MDL Number MFCD01631579
Beilstein 2525206
Synonym 1-Bromo-4-methoxy-3-(trifluoromethyl)benzene; 3-Bromo-6-methoxybenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2443604

 
alfa aesar™
B2443604
2g Each for $107.30

AAB2443609

 
alfa aesar™
B2443609
10g Each for $357.11
Description & Specifications

Specifications

Formula Weight 255.04
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)anisole
Assay 97%
Molecular Formula C8H6BrF3O
MDL Number MFCD01631579
CAS 1514-11-0
Beilstein 2525206
Synonym 1-Bromo-4-methoxy-3-(trifluoromethyl)benzene; 3-Bromo-6-methoxybenzotrifluoride
Boiling Point 120°C (30mmHg)
Quantity 2g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.