Please login to your online account to display your discounted pricing

4-Bromo-2-thiophenecarboxylic acid, 97%, Maybridge


Quantity 25g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC28501EB

 View more versions of this product

Catalog No. CC28501EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 207.05
Quantity 25g
Assay 95%
Chemical Name or Material 4-Bromo-2-thiophenecarboxylic acid
Packaging Amber Glass Bottle
Molecular Formula C5H3BrO2S
CAS 16694-18-1
Assay Percent Range 95%

Specifications

Chemical Name or Material 4-Bromo-2-thiophenecarboxylic acid
Assay 95%
Assay Percent Range 95%
Molecular Formula C5H3BrO2S
Formula Weight 207.05