missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Bromo-2-thienyl)methanol, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC28509DA

 View more versions of this product

Catalog No. CC28509DA



Specifications

Specifications

(4-Bromo-2-thienyl)methanol
1g
96.5
97%
MFCD04115392
PXZNJHHUYJRFPZ-UHFFFAOYSA-N
(4-bromothiophen-2-yl)methanol
2795484
97%
Amber Glass Bottle
79757-77-0
100.0
C5H5BrOS
2-thiophenemethanol, 4-bromo, 2-thiophenemethanol,4-bromo, 3-bromo-5-thienyl methanol, 4-bromo-2-hydroxymethyl thiophene, 4-bromo-2-hydroxymethylthiophene, 4-bromo-2-thienyl methanol, 4-bromo-2-thiophenemethanol, 4-bromo-thiophene-2-methanol, 4-bromothiophen-2-yl methanol, acmc-1bm1t
C1=C(SC=C1Br)CO
193.058
193.06

Chemical Identifiers

79757-77-0
193.058
PXZNJHHUYJRFPZ-UHFFFAOYSA-N
2795484
C1=C(SC=C1Br)CO
C5H5BrOS
MFCD04115392
2-thiophenemethanol, 4-bromo, 2-thiophenemethanol,4-bromo, 3-bromo-5-thienyl methanol, 4-bromo-2-hydroxymethyl thiophene, 4-bromo-2-hydroxymethylthiophene, 4-bromo-2-thienyl methanol, 4-bromo-2-thiophenemethanol, 4-bromo-thiophene-2-methanol, 4-bromothiophen-2-yl methanol, acmc-1bm1t
(4-bromothiophen-2-yl)methanol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents