Please login to your online account to display your discounted pricing

4-Bromo-2-nitrotoluene, 98%, ACROS Organics™

$103.09 - $103.09

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-nitrotoluene
CAS 60956-26-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6BrNO2
Formula Weight 216.03
MDL Number MFCD00041243
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC276210010

  SDS
thermo fisher scientific
AC276210010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC276210050

  SDS
thermo fisher scientific
276210050
Glass bottle 5g Each for $103.09
Description & Specifications

Specifications

Chemical Name or Material 4-Bromo-2-nitrotoluene
CAS 60956-26-5
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 43° to 47°C
Color Yellow to light orange
Physical Form Crystals or Crystalline Powder
Quantity 1g
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6BrNO2
Formula Weight 216.03
MDL Number MFCD00041243
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.