missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-2-nitrotoluene, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC276210010
Specifications
4-Bromo-2-nitrotoluene | |
Authentic | |
130.0°C (12.0 mmHg) | |
43.0°C to 46.0°C | |
60956-26-5 | |
100.0 | |
C7H6BrNO2 | |
2-nitro-4-bromotoluene, 4-brom-2-nitrotoluol, 4-bromo-1-methyl-2-nitro-benzene, 4-bromo-2-nitrotoluene, 4-bromo-6-nitrotoluene, 5-bromo-2-methylnitrobenzene, benzene, 4-bromo-1-methyl-2-nitro, bromo 4--2-nitrotoluene, maybridge1_002072, pubchem10403 | |
KZNXALJXBRSMFL-UHFFFAOYSA-N | |
4-bromo-1-methyl-2-nitrobenzene | |
123546 | |
Crystals or Crystalline Powder |
>110°C | |
Glass bottle | |
Orange to Yellow | |
1g | |
97.5 | |
97.5% min. (GC) | |
MFCD00041243 | |
Solubility in water: insoluble. | |
CC1=C(C=C(C=C1)Br)[N+](=O)[O-] | |
216.03 | |
216.03 | |
98% |
Chemical Identifiers
60956-26-5 | |
216.03 | |
KZNXALJXBRSMFL-UHFFFAOYSA-N | |
123546 | |
CC1=C(C=C(C=C1)Br)[N+](=O)[O-] |
C7H6BrNO2 | |
MFCD00041243 | |
2-nitro-4-bromotoluene, 4-brom-2-nitrotoluol, 4-bromo-1-methyl-2-nitro-benzene, 4-bromo-2-nitrotoluene, 4-bromo-6-nitrotoluene, 5-bromo-2-methylnitrobenzene, benzene, 4-bromo-1-methyl-2-nitro, bromo 4--2-nitrotoluene, maybridge1_002072, pubchem10403 | |
4-bromo-1-methyl-2-nitrobenzene |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 262-536-3