Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-nitrophenol, 98%

$226.01 - $226.01

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-nitrophenol
CAS 7693-52-9
Molecular Formula C6H4BrNO3
Formula Weight 218
MDL Number MFCD00082540
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2652914

  SDS
alfa aesar™
H2652914
25g Each for $226.01
Description & Specifications

Specifications

Formula Weight 218
Chemical Name or Material 4-Bromo-2-nitrophenol
Melting Point 88° to 91°C
Quantity 25g
Assay 98%
Molecular Formula C6H4BrNO3
MDL Number MFCD00082540
CAS 7693-52-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SJ8370000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.