missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-nitroanisole, 97%

Manufacturer:  Alfa Aesar™ H6487722

 View more versions of this product

Catalog No. AAH6487722



Specifications

Specifications

85°C to 100°C
33696-00-3
MFCD00055529
1-methoxy-2-nitro-4-bromobenzene, 2-methoxy-5-bromonitrobenzene, 3-nitro-4-methoxy bromobenzene, 4-brom-2-nitroanisol, 4-brom-2-nitroanisole, 4-bromo-2-nitro anisole, 4-bromo-2-nitroanisole, 5-bromo-2-methoxynitrobenzene, benzene, 4-bromo-1-methoxy-2-nitro, pubchem5341
COC1=C(C=C(C=C1)Br)[N+](=O)[O-]
232.033
232.03
97%
100g
C7H6BrNO3
2259903
ORBHQHXVVMZIDP-UHFFFAOYSA-N
4-bromo-1-methoxy-2-nitrobenzene
118533
Solid

Chemical Identifiers

33696-00-3
232.033
ORBHQHXVVMZIDP-UHFFFAOYSA-N
118533
COC1=C(C=C(C=C1)Br)[N+](=O)[O-]
C7H6BrNO3
MFCD00055529
1-methoxy-2-nitro-4-bromobenzene, 2-methoxy-5-bromonitrobenzene, 3-nitro-4-methoxy bromobenzene, 4-brom-2-nitroanisol, 4-brom-2-nitroanisole, 4-bromo-2-nitro anisole, 4-bromo-2-nitroanisole, 5-bromo-2-methoxynitrobenzene, benzene, 4-bromo-1-methoxy-2-nitro, pubchem5341
4-bromo-1-methoxy-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 251-642-5

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents