Learn More
4-Bromo-2-nitroanisole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6487714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Bromo-2-nitroanisole | |
85°C to 100°C | |
MFCD00055529 | |
2259903 | |
ORBHQHXVVMZIDP-UHFFFAOYSA-N | |
4-bromo-1-methoxy-2-nitrobenzene | |
118533 | |
97% |
33696-00-3 | |
C7H6BrNO3 | |
25 g | |
4-bromo-2-nitroanisole, 2-methoxy-5-bromonitrobenzene, 4-brom-2-nitroanisole, 4-bromo-2-nitro anisole, 5-bromo-2-methoxynitrobenzene, 3-nitro-4-methoxy bromobenzene, 1-methoxy-2-nitro-4-bromobenzene, benzene, 4-bromo-1-methoxy-2-nitro, pubchem5341, 4-brom-2-nitroanisol | |
COC1=C(C=C(C=C1)Br)[N+](=O)[O-] | |
232.033 | |
232.03 | |
Solid |
Chemical Identifiers
33696-00-3 | |
232.033 | |
ORBHQHXVVMZIDP-UHFFFAOYSA-N | |
118533 | |
COC1=C(C=C(C=C1)Br)[N+](=O)[O-] |
C7H6BrNO3 | |
MFCD00055529 | |
4-bromo-2-nitroanisole, 2-methoxy-5-bromonitrobenzene, 4-brom-2-nitroanisole, 4-bromo-2-nitro anisole, 5-bromo-2-methoxynitrobenzene, 3-nitro-4-methoxy bromobenzene, 1-methoxy-2-nitro-4-bromobenzene, benzene, 4-bromo-1-methoxy-2-nitro, pubchem5341, 4-brom-2-nitroanisol | |
4-bromo-1-methoxy-2-nitrobenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 251-642-5
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only