missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-nitro-6-(trifluoromethyl)aniline, 98%

Manufacturer:  Alfa Aesar™ H6193306

 View more versions of this product

Catalog No. AAH6193306



Specifications

Specifications

4-Bromo-2-nitro-6-(trifluoromethyl)aniline
157026-18-1
MFCD09835318
JWIAFKYOENFPNQ-UHFFFAOYSA-N
4-bromo-2-nitro-6-(trifluoromethyl)aniline
10850672
Solid
5g
C7H4BrF3N2O2
2-amino-5-bromo-3-nitrobenzotrifluoride
C1=C(C=C(C(=C1[N+](=O)[O-])N)C(F)(F)F)Br
285.02
285.02
98%

Chemical Identifiers

157026-18-1
285.02
JWIAFKYOENFPNQ-UHFFFAOYSA-N
10850672
C1=C(C=C(C(=C1[N+](=O)[O-])N)C(F)(F)F)Br
C7H4BrF3N2O2
MFCD09835318
2-amino-5-bromo-3-nitrobenzotrifluoride
4-bromo-2-nitro-6-(trifluoromethyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents