missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-2-(methylthio)pyrimidine, 97%

Manufacturer:  Alfa Aesar™ H6412406

 View more versions of this product

Catalog No. AAH6412406



Specifications

Specifications

4-Bromo-2-(methylthio)pyrimidine
30°C to 40°C
959236-97-6
MFCD09878932
21913366
TZSQAGYDWPPKPE-UHFFFAOYSA-N
4-bromo-2-methylsulfanylpyrimidine
45787531
Low Melting Solid
Brown to Yellow
5g
C5H5BrN2S
UN1759
4-bromo-2-methylthio pyrimidine, 4-bromo-2-methylsulfanyl pyrimidine, pyrimidine, 4-bromo-2-methylthio, 4-bromo-2-methylsulfanyl-pyrimidine, 2-methylthio-4-bromopyrimidine, 2-methylthio-4-bromopyrimidine
CSC1=NC=CC(=N1)Br
205.073
205.07
97%

Chemical Identifiers

959236-97-6
205.073
TZSQAGYDWPPKPE-UHFFFAOYSA-N
45787531
CSC1=NC=CC(=N1)Br
C5H5BrN2S
MFCD09878932
4-bromo-2-methylthio pyrimidine, 4-bromo-2-methylsulfanyl pyrimidine, pyrimidine, 4-bromo-2-methylthio, 4-bromo-2-methylsulfanyl-pyrimidine, 2-methylthio-4-bromopyrimidine, 2-methylthio-4-bromopyrimidine
4-bromo-2-methylsulfanylpyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

storageNote1 : -30° to -10°C

SDS Documents